missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methyl-2-butanone 95.0+%, TCI America™

Catalog No. p-7130553
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H047125ML 25 mL
H0471100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H047125ML Supplier TCI America Supplier No. H047125ML
Only null left

Chemical Identifiers

CAS 115-22-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004460
InChI Key BNDRWEVUODOUDW-UHFFFAOYSA-N
Synonym 3-hydroxy-3-methyl-2-butanone, 3-methylacetoin, methylacetoin, 2-butanone, 3-hydroxy-3-methyl, dimethylacetylcarbinol, 2-hydroxy-2-methyl-3-butanone, 2-methyl-2-hydroxybutan-3-one, 1-hydroxy-1-methylethyl methyl ketone, 3-hydroxy-3-methyl-butan-2-one, 3-hydroxy-3-ethylbutanone
PubChem CID 8261
IUPAC Name 3-hydroxy-3-methylbutan-2-one
SMILES CC(=O)C(C)(C)O

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 102.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone
EINECSNumber (2)-0594
TSCA Yes
Recommended Storage Refrigerator