accessibility menu, dialog, popup

UserName

CAS RN 115-22-0

IUPAC Name: 3-hydroxy-3-methylbutan-2-one
Synonyms: 3-hydroxy-3-methyl-2-butanone
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: BNDRWEVUODOUDW-UHFFFAOYSA-N
SMILES: CC(=O)C(C)(C)O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)