missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methyl-2-butanone, 90+%

Catalog No. p-7058989
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4372414 25 g
AA4372422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4372414 Supplier Thermo Scientific Chemicals Supplier No. 04372414
Only null left

CAS: 115-22-0 | C5H10O2 | 102.133 g/mol

3-Hydroxy-3-methyl-2-butanone used to produce 2,5-dihydroxy-2-methyl-heptan-3-one, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115-22-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004460
InChI Key BNDRWEVUODOUDW-UHFFFAOYSA-N
Synonym 3-hydroxy-3-methyl-2-butanone, 3-methylacetoin, methylacetoin, 2-butanone, 3-hydroxy-3-methyl, dimethylacetylcarbinol, 2-hydroxy-2-methyl-3-butanone, 2-methyl-2-hydroxybutan-3-one, 1-hydroxy-1-methylethyl methyl ketone, 3-hydroxy-3-methyl-butan-2-one, 3-hydroxy-3-ethylbutanone
PubChem CID 8261
IUPAC Name 3-hydroxy-3-methylbutan-2-one
SMILES CC(=O)C(C)(C)O

Specifications

Density 0.96 g/mL
Boiling Point 140°C to 141°C
Flash Point 42°C (107.6°F)
Refractive Index 1.415
Quantity 25 g
UN Number UN1224
Solubility Information Soluble in chloroform.
Formula Weight 102.13
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 204-073-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only