missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-pyridinecarboxylic Acid 98.0+%, TCI America™

$179.05 - $710.39

Chemical Identifiers

CAS 874-24-8
Molecular Formula C6H4NO3
Molecular Weight (g/mol) 138.10
MDL Number MFCD00006294
InChI Key BRARRAHGNDUELT-UHFFFAOYSA-M
Synonym 3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
PubChem CID 13401
ChEBI CHEBI:64342
IUPAC Name 3-hydroxypyridine-2-carboxylate
SMILES OC1=CC=CN=C1C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H07875G
View Documents
TCI America
H07875G
5 g N/A
Only null left
 
H078725G
View Documents
TCI America
H078725G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

874-24-8
138.10
BRARRAHGNDUELT-UHFFFAOYSA-M
13401
3-hydroxypyridine-2-carboxylate
C6H4NO3
MFCD00006294
3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
CHEBI:64342
OC1=CC=CN=C1C([O-])=O

Specifications

874-24-8
White-Yellow
MFCD00006294
3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
OC1=CC=CN=C1C([O-])=O
138.10
CHEBI:64342
≥98.0% (T)
3-Hydroxy-2-pyridinecarboxylic Acid
204°C
C6H4NO3
5 g
BRARRAHGNDUELT-UHFFFAOYSA-M
3-hydroxypyridine-2-carboxylate
13401
139.11
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents