missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-2-nitrobenzoic Acid 98.0+%, TCI America™
$111.66 - $519.80
Chemical Identifiers
| CAS | 602-00-6 |
|---|---|
| Molecular Formula | C7H5NO5 |
| Molecular Weight (g/mol) | 183.119 |
| MDL Number | MFCD00017003 |
| InChI Key | KPDBKQKRDJPBRM-UHFFFAOYSA-N |
| Synonym | 3-hydroxy-2-nitrobenzoic acid, 3-hydroxy-2-nitrobenzoicacid, 3-hydroxy-2-nitro-benzoic acid, 2-nitro-3-hydroxybenzoic acid, pubchem8846, nitro-m-oxybenzoesaure, benzoic acid, hydroxynitro, ksc288q8j, acmc-1b214, 2-nitro-3-oxidanyl-benzoic acid |
| PubChem CID | 293291 |
| SMILES | C1=CC(=C(C(=C1)O)[N+](=O)[O-])C(=O)O |
Chemical Identifiers
| 602-00-6 | |
| 183.119 | |
| KPDBKQKRDJPBRM-UHFFFAOYSA-N | |
| 293291 |
| C7H5NO5 | |
| MFCD00017003 | |
| 3-hydroxy-2-nitrobenzoic acid, 3-hydroxy-2-nitrobenzoicacid, 3-hydroxy-2-nitro-benzoic acid, 2-nitro-3-hydroxybenzoic acid, pubchem8846, nitro-m-oxybenzoesaure, benzoic acid, hydroxynitro, ksc288q8j, acmc-1b214, 2-nitro-3-oxidanyl-benzoic acid | |
| C1=CC(=C(C(=C1)O)[N+](=O)[O-])C(=O)O |
Specifications
| 602-00-6 | |
| C7H5NO5 | |
| 1 g | |
| KPDBKQKRDJPBRM-UHFFFAOYSA-N | |
| 183.119 | |
| 183.12 | |
| Crystalline Powder |
| 182°C | |
| MFCD00017003 | |
| 3-hydroxy-2-nitrobenzoic acid, 3-hydroxy-2-nitrobenzoicacid, 3-hydroxy-2-nitro-benzoic acid, 2-nitro-3-hydroxybenzoic acid, pubchem8846, nitro-m-oxybenzoesaure, benzoic acid, hydroxynitro, ksc288q8j, acmc-1b214, 2-nitro-3-oxidanyl-benzoic acid | |
| C1=CC(=C(C(=C1)O)[N+](=O)[O-])C(=O)O | |
| 293291 | |
| ≥98.0% (GC,T) | |
| 3-Hydroxy-2-nitrobenzoic Acid |
Safety and Handling
TSCA : No