missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. H13245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13245G 5 g
H13241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13245G Supplier TCI America Supplier No. H13245G

Chemical Identifiers

CAS 602-00-6
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00017003
InChI Key KPDBKQKRDJPBRM-UHFFFAOYSA-N
Synonym 3-hydroxy-2-nitrobenzoic acid, 3-hydroxy-2-nitrobenzoicacid, 3-hydroxy-2-nitro-benzoic acid, 2-nitro-3-hydroxybenzoic acid, pubchem8846, nitro-m-oxybenzoesaure, benzoic acid, hydroxynitro, ksc288q8j, acmc-1b214, 2-nitro-3-oxidanyl-benzoic acid
PubChem CID 293291
SMILES C1=CC(=C(C(=C1)O)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 182°C
Quantity 5 g
Formula Weight 183.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-nitrobenzoic Acid