missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-nitrobenzoic acid, 98+%

Catalog No. p-7046022
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1284104 2 g
AAL1284109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1284104 Supplier Thermo Scientific Chemicals Supplier No. L1284104

CAS: 602-00-6 | C7H5NO5 | 183.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 602-00-6
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00017003
InChI Key KPDBKQKRDJPBRM-UHFFFAOYSA-N
Synonym 3-hydroxy-2-nitrobenzoic acid, 3-hydroxy-2-nitrobenzoicacid, 3-hydroxy-2-nitro-benzoic acid, 2-nitro-3-hydroxybenzoic acid, pubchem8846, nitro-m-oxybenzoesaure, benzoic acid, hydroxynitro, ksc288q8j, acmc-1b214, 2-nitro-3-oxidanyl-benzoic acid
PubChem CID 293291
SMILES C1=CC(=C(C(=C1)O)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 177°C to 181°C
Quantity 2 g
Beilstein 2372248
Formula Weight 183.12
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxy-2-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only