missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthoic Acid 98.0+%, TCI America™

Catalog No. H028125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H028125G 25 g
H0281500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H028125G Supplier TCI America Supplier No. H028125G

Chemical Identifiers

CAS 92-70-6
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00004103
InChI Key ALKYHXVLJMQRLQ-UHFFFAOYSA-N
Synonym 3-hydroxy-2-naphthoic acid, 2-hydroxy-3-naphthoic acid, bon acid, bona, developer bon, 2-naphthol-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy, b.o.n. acid, c.i. developer 20, miketazol developer ons
PubChem CID 7104
ChEBI CHEBI:80383
IUPAC Name 3-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)O

Specifications

Melting Point 220°C
Color Yellow
Quantity 25 g
Formula Weight 188.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-naphthoic Acid
EINECSNumber (4)-0398
RTECSNumber QL1755000
TSCA Yes