missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthoic acid hydrazide, 98%

Catalog No. AAA1619214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1619214 25 g
AAA1619222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1619214 Supplier Thermo Scientific Chemicals Supplier No. A1619214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5341-58-2 | C11H10N2O2 | 202.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5341-58-2
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD00004097
InChI Key FDNAQCWUERCJBK-UHFFFAOYSA-N
Synonym 3-hydroxy-2-naphthohydrazide, 3-hydroxy-2-naphthoic acid hydrazide, 2-hydroxy-3-naphthoic acid hydrazide, 2-naphthalenecarboxylic acid, 3-hydroxy-, hydrazide, 3-hydroxy-2-naphthoylhydrazine, 2-hydroxy-3-naphthoylhydrazine, 2-naphthoic acid, 3-hydroxy-, hydrazide, 3-hydroxy-2-naphthoic hydrazide, beta-hydroxynaphthoic hydrazide, 3-???naphthalen-2-ol
PubChem CID 72699
IUPAC Name 3-hydroxynaphthalene-2-carbohydrazide
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)NN)O

Specifications

Melting Point ∼205°C (decomposition)
Quantity 25 g
Beilstein 395153
Formula Weight 202.21
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-2-naphthoic acid hydrazide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 226-282-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only