missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthoic acid, 98%

Catalog No. AAA1921430
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1921430 250 g
AAA192140B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1921430 Supplier Thermo Scientific Chemicals Supplier No. A1921430

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92-70-6 | C11H8O3 | 188.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-70-6
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00004103
InChI Key ALKYHXVLJMQRLQ-UHFFFAOYSA-N
Synonym 3-hydroxy-2-naphthoic acid, 2-hydroxy-3-naphthoic acid, bon acid, bona, developer bon, 2-naphthol-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy, b.o.n. acid, c.i. developer 20, miketazol developer ons
PubChem CID 7104
ChEBI CHEBI:80383
IUPAC Name 3-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)O

Specifications

Melting Point 218°C to 222°C
Flash Point 220°C (428°F)
Quantity 250 g
Beilstein 744100
Merck Index 14,4835
Formula Weight 188.18
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-2-naphthoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-180-8
RTECSNumber QL1755000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only