missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-2-methyl-4-pyrone 99.0+%, TCI America™
$57.79 - $505.16
Chemical Identifiers
| CAS | 118-71-8 |
|---|---|
| Molecular Formula | C6H6O3 |
| Molecular Weight (g/mol) | 126.11 |
| MDL Number | MFCD00006578 |
| InChI Key | XPCTZQVDEJYUGT-UHFFFAOYSA-N |
| Synonym | maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl |
| PubChem CID | 8369 |
| ChEBI | CHEBI:69438 |
| IUPAC Name | 3-hydroxy-2-methyl-4H-pyran-4-one |
| SMILES | CC1=C(O)C(=O)C=CO1 |
Chemical Identifiers
| 118-71-8 | |
| 126.11 | |
| XPCTZQVDEJYUGT-UHFFFAOYSA-N | |
| 8369 | |
| 3-hydroxy-2-methyl-4H-pyran-4-one |
| C6H6O3 | |
| MFCD00006578 | |
| maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl | |
| CHEBI:69438 | |
| CC1=C(O)C(=O)C=CO1 |
Specifications
| 118-71-8 | |
| White | |
| MFCD00006578 | |
| maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl | |
| CC1=C(O)C(=O)C=CO1 | |
| 126.11 | |
| CHEBI:69438 | |
| ≥99.0% (HPLC,T) | |
| 3-Hydroxy-2-methyl-4-pyrone |
| 162°C | |
| C6H6O3 | |
| 25 g | |
| XPCTZQVDEJYUGT-UHFFFAOYSA-N | |
| 3-hydroxy-2-methyl-4H-pyran-4-one | |
| 8369 | |
| 126.11 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3642
RTECSNumber : UQ1050000
TSCA : Yes