missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methyl-4-pyrone 99.0+%, TCI America™

Supplier:  TCI America M0673100G

 View more versions of this product

Catalog No. M0673100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Hydroxy-2-methyl-4-pyrone
162°C
C6H6O3
100 g
XPCTZQVDEJYUGT-UHFFFAOYSA-N
3-hydroxy-2-methyl-4H-pyran-4-one
8369
126.11
Crystalline Powder
118-71-8
White
MFCD00006578
maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl
CC1=C(O)C(=O)C=CO1
126.11
CHEBI:69438
≥99.0% (HPLC,T)

Chemical Identifiers

118-71-8
126.11
XPCTZQVDEJYUGT-UHFFFAOYSA-N
8369
3-hydroxy-2-methyl-4H-pyran-4-one
C6H6O3
MFCD00006578
maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl
CHEBI:69438
CC1=C(O)C(=O)C=CO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3642

RTECSNumber : UQ1050000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions