missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2'-methoxy-2-naphthanilide 98.0+%, TCI America™

Catalog No. H031725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H031725G 25 g
H0317500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H031725G Supplier TCI America Supplier No. H031725G

Chemical Identifiers

CAS 135-62-6
Molecular Formula C18H14NNaO3
Molecular Weight (g/mol) 315.30
MDL Number MFCD00021630
InChI Key OXXQDUUQLWQUEX-UHFFFAOYSA-M
Synonym 2-Hydroxy-3-naphthoic Acid o-Anisidide, Naphthol AS-OL, 3-Hydroxy-N-(2-methoxyphenyl)-2-naphthamide, Azoic Coupling Component 20
PubChem CID 67274
IUPAC Name sodium 3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-olate
SMILES [Na+].COC1=CC=CC=C1NC(=O)C1=C([O-])C=C2C=CC=CC2=C1

Specifications

Melting Point 162°C
Color Yellow
Quantity 25 g
Formula Weight 293.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2′-methoxy-2-naphthanilide
EINECSNumber (5)-2288
TSCA Yes