missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2,2-dimethylpropionic acid, 97+%

Catalog No. AAL1285818
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1285818 50 g
AAL1285830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1285818 Supplier Thermo Scientific Chemicals Supplier No. L1285818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4835-90-9 | C5H10O3 | 118.132 g/mol

3-Hydroxy-2,2-dimethylpropionic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-2,2-dimethylpropionic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4835-90-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00059953
InChI Key RDFQSFOGKVZWKF-UHFFFAOYSA-N
Synonym hydroxypivalic acid, 3-hydroxypivalic acid, 2,2-dimethyl-3-hydroxypropionic acid, propanoic acid, 3-hydroxy-2,2-dimethyl, 3-hydroxy-2,2-dimethylpropionic acid, unii-bjp2cxk56p, bjp2cxk56p, acmc-1arbc, beta-hydroxy pivalic acid
PubChem CID 78548
IUPAC Name 3-hydroxy-2,2-dimethylpropanoic acid
SMILES CC(C)(CO)C(=O)O

Specifications

Melting Point 122°C to 126°C
Odor Odorless
Quantity 50 g
Beilstein 1747025
Solubility Information Soluble in water.
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-2,2-dimethylpropionic acid
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 225-419-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only