missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2,2,4-trimethylpentyl Isobutyrate 60.0+%, TCI America™

Catalog No. p-7131390
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I040525ML 25 mL
I0405500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I040525ML Supplier TCI America Supplier No. I040525ML
Only null left

(contains ca. 40% 2,2,4-Trimethyl-1,3-pentanediol 3-Monoisobutyrate)

Chemical Identifiers

CAS 25265-77-4
Molecular Formula C12H24O3
Molecular Weight (g/mol) 216.321
MDL Number MFCD00148967
InChI Key DAFHKNAQFPVRKR-UHFFFAOYSA-N
Synonym texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
PubChem CID 6490
IUPAC Name (3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate
SMILES CC(C)C(C(C)(C)COC(=O)C(C)C)O

Specifications

Melting Point -50°C
Color Yellow
Boiling Point 253°C
Quantity 25 mL
Formula Weight 216.32
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-2,2,4-trimethylpentyl Isobutyrate (contains ca. 40% 2,2,4-Trimethyl-1,3-pentanediol 3-Mono
EINECSNumber (2)-0778
RTECSNumber UF6000000
TSCA Yes