missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1-methacryloyloxyadamantane 98.0+%, TCI America™

Catalog No. H15435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15435G 5 g
H154325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15435G Supplier TCI America Supplier No. H15435G

Chemical Identifiers

CAS 115372-36-6
Molecular Formula C14H20O3
Molecular Weight (g/mol) 236.311
MDL Number MFCD13152044
InChI Key OOIBFPKQHULHSQ-UHFFFAOYSA-N
Synonym 3-Hydroxy-1-adamantyl Methacrylate, Methacrylic Acid 3-Hydroxy-1-adamantyl Ester
PubChem CID 11856523
IUPAC Name (3-hydroxy-1-adamantyl) 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC12CC3CC(C1)CC(C3)(C2)O

Specifications

Melting Point 91°C
Color White
Quantity 5 g
Formula Weight 236.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-1-methacryloyloxyadamantane
EINECSNumber (4)-1878
TSCA Yes