missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hexyn-1-ol 97.0+%, TCI America™
Supplier: TCI America H046025ML
Specifications
| 3-Hexyn-1-ol | |
| Yellow | |
| C6H10O | |
| 25 mL | |
| ODEHKVYXWLXRRR-UHFFFAOYSA-N | |
| hex-3-yn-1-ol | |
| 66083 | |
| ≥97.0% (GC) |
| 1002-28-4 | |
| 161°C | |
| MFCD00002957 | |
| 3-hexyn-1-ol, 3-hexynol, 3-hexyne-1-ol, unii-x07kyi4gth, x07kyi4gth, acmc-2097nm, 3-hexyn-1-ol, 98 %, 3-hexyn-1-ol 5ml | |
| CCC#CCCO | |
| 98.145 | |
| 98.15 | |
| Liquid |
Chemical Identifiers
| 1002-28-4 | |
| 98.145 | |
| ODEHKVYXWLXRRR-UHFFFAOYSA-N | |
| 66083 | |
| CCC#CCCO |
| C6H10O | |
| MFCD00002957 | |
| 3-hexyn-1-ol, 3-hexynol, 3-hexyne-1-ol, unii-x07kyi4gth, x07kyi4gth, acmc-2097nm, 3-hexyn-1-ol, 98 %, 3-hexyn-1-ol 5ml | |
| hex-3-yn-1-ol |
Safety and Handling
EINECSNumber : (2)-3191
TSCA : Yes