missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyn-1-ol 97.0+%, TCI America™

Catalog No. p-7130546
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04605ML 5 mL
H046025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04605ML Supplier TCI America Supplier No. H04605ML
Only null left

Chemical Identifiers

CAS 1002-28-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00002957
InChI Key ODEHKVYXWLXRRR-UHFFFAOYSA-N
Synonym 3-hexyn-1-ol, 3-hexynol, 3-hexyne-1-ol, unii-x07kyi4gth, x07kyi4gth, acmc-2097nm, 3-hexyn-1-ol, 98 %, 3-hexyn-1-ol 5ml
PubChem CID 66083
IUPAC Name hex-3-yn-1-ol
SMILES CCC#CCCO

Specifications

Color Yellow
Boiling Point 161°C
Quantity 5 mL
Formula Weight 98.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hexyn-1-ol
EINECSNumber (2)-3191
TSCA Yes