missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hexylthiophene 98.0+%, TCI America™
$61.51 - $546.17
Chemical Identifiers
| CAS | 1693-86-3 |
|---|---|
| Molecular Formula | C10H16S |
| Molecular Weight (g/mol) | 168.298 |
| MDL Number | MFCD00143181 |
| InChI Key | JEDHEMYZURJGRQ-UHFFFAOYSA-N |
| Synonym | 3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+% |
| PubChem CID | 566849 |
| IUPAC Name | 3-hexylthiophene |
| SMILES | CCCCCCC1=CSC=C1 |
Chemical Identifiers
| 1693-86-3 | |
| 168.298 | |
| JEDHEMYZURJGRQ-UHFFFAOYSA-N | |
| 566849 | |
| CCCCCCC1=CSC=C1 |
| C10H16S | |
| MFCD00143181 | |
| 3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+% | |
| 3-hexylthiophene |
Specifications
| 1693-86-3 | |
| 103°C | |
| MFCD00143181 | |
| 1993 | |
| JEDHEMYZURJGRQ-UHFFFAOYSA-N | |
| 3-hexylthiophene | |
| 566849 | |
| ≥98.0% (GC) | |
| 3-Hexylthiophene |
| Colorless | |
| C10H16S | |
| 1 g | |
| 3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+% | |
| CCCCCCC1=CSC=C1 | |
| 168.298 | |
| 168.30 | |
| Liquid |
Safety and Handling
TSCA : No