accessibility menu, dialog, popup

UserName

H3CS

CAS RN 1693-86-3

IUPAC Name: 3-hexylthiophene
Synonyms: 3-hexylthiophene
Molecular Weight (g/mol): 168.3
Molecular Formula: C10H16S
InChi Key: JEDHEMYZURJGRQ-UHFFFAOYSA-N
SMILES: CCCCCCC1=CSC=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (8)
  • (2)
  • (3)
  • (3)