missing translation for 'onlineSavingsMsg'
Learn More

3-Hexylthiophene 98.0+%, TCI America™

Catalog No. H07561G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07561G 1 g
H07565G 5 g
H075625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H07561G Supplier TCI America Supplier No. H07561G

Chemical Identifiers

CAS 1693-86-3
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.298
MDL Number MFCD00143181
InChI Key JEDHEMYZURJGRQ-UHFFFAOYSA-N
Synonym 3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
PubChem CID 566849
IUPAC Name 3-hexylthiophene
SMILES CCCCCCC1=CSC=C1

Specifications

Color Colorless
Boiling Point 103°C
Quantity 1 g
UN Number 1993
Formula Weight 168.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hexylthiophene