missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hexanol 98.0+%, TCI America™
Supplier: TCI America H013225ML
Specifications
| 3-Hexanol | |
| Colorless | |
| C6H14O | |
| 25 mL | |
| 3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d | |
| CCCC(CC)O | |
| 102.177 | |
| 102.18 | |
| Liquid |
| 623-37-0 | |
| 135°C | |
| MFCD00004582 | |
| 2282 | |
| ZOCHHNOQQHDWHG-UHFFFAOYSA-N | |
| hexan-3-ol | |
| 12178 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 623-37-0 | |
| 102.177 | |
| ZOCHHNOQQHDWHG-UHFFFAOYSA-N | |
| 12178 | |
| CCCC(CC)O |
| C6H14O | |
| MFCD00004582 | |
| 3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d | |
| hexan-3-ol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : MIO857000
TSCA : Yes