accessibility menu, dialog, popup

UserName

H3COHCH3

CAS RN 623-37-0

IUPAC Name: hexan-3-ol
Synonyms: 3-hexanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: ZOCHHNOQQHDWHG-UHFFFAOYNA-N
SMILES: CCCC(O)CC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)