missing translation for 'onlineSavingsMsg'
Learn More

3-Hexadecylthiophene 97.0+%, TCI America™

Catalog No. H14621G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14621G 1 g
H14625G 5 g
H146225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H14621G Supplier TCI America Supplier No. H14621G

Chemical Identifiers

CAS 119269-24-8
Molecular Formula C20H36S
Molecular Weight (g/mol) 308.57
MDL Number MFCD07368973
InChI Key FRVZSODZVJPMKO-UHFFFAOYSA-N
Synonym 3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275
PubChem CID 2769638
IUPAC Name 3-hexadecylthiophene
SMILES CCCCCCCCCCCCCCCCC1=CSC=C1

Specifications

Melting Point 33°C
Color White-Yellow
Quantity 1 g
Formula Weight 308.57
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Hexadecylthiophene