missing translation for 'onlineSavingsMsg'
Learn More

3-Heptenoic Acid 90.0+%, TCI America™

Catalog No. H04275ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04275ML 5 mL
H042725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04275ML Supplier TCI America Supplier No. H04275ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29901-85-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00058999
InChI Key BCYXFRMDZWKZSF-PLNGDYQASA-N
PubChem CID 6434067
IUPAC Name (Z)-hept-3-enoic acid
SMILES CCCC=CCC(=O)O

Specifications

Color Yellow
Boiling Point 228°C
Quantity 5 mL
UN Number 3265
Formula Weight 128.17
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Heptenoic Acid
EINECSNumber (2)-0609
TSCA No