accessibility menu, dialog, popup

UserName

CAS RN 29901-85-7

IUPAC Name: (3Z)-hept-3-enoic acid
Synonyms: (3Z)-hept-3-enoic acid
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: BCYXFRMDZWKZSF-PLNGDYQASA-N
SMILES: CCC\C=C/CC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results