missing translation for 'onlineSavingsMsg'
Learn More

3-Heptenoic Acid 90.0+%, TCI America™

Supplier:  TCI America H042725ML

Quantity 25 mL

 View more versions of this product

Catalog No. H042725ML


Only null left

Description

Description

Specifications

Chemical Identifiers

29901-85-7
128.171
BCYXFRMDZWKZSF-PLNGDYQASA-N
(Z)-hept-3-enoic acid
C7H12O2
MFCD00058999
6434067
CCCC=CCC(=O)O

Specifications

3-Heptenoic Acid
Yellow
C7H12O2
MFCD00058999
BCYXFRMDZWKZSF-PLNGDYQASA-N
(Z)-hept-3-enoic acid
6434067
≥90.0% (GC)
29901-85-7
228°C
25 mL
3265
CCCC=CCC(=O)O
128.171
128.17
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0609

TSCA : No

SDS
Documents

Documents

Promotions

Promotions