missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanol 98.0+%, TCI America™

Catalog No. p-7130301
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
H003525ML 25 mL
H0035500ML 500 mL
2 options

Catalog No. H003525ML

Supplier: TCI America H003525ML

Only null left

Chemical Identifiers

CAS 589-82-2
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00004586
InChI Key RZKSECIXORKHQS-UHFFFAOYSA-N
Synonym 3-heptanol, 3-hydroxyheptane, ethyl butyl carbinol, butyl ethyl carbinol, butylethylcarbinol, xi-3-heptanol, fema no. 3547, ethylbutylcarbinol, heptanol-3, 3-heptyl alcohol
PubChem CID 11520
IUPAC Name heptan-3-ol
SMILES CCCCC(CC)O

Specifications

Color Colorless
Boiling Point 156°C
Quantity 25 mL
UN Number 1987
Formula Weight 116.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Heptanol
EINECSNumber (2)-0217
RTECSNumber MJ3150000
TSCA Yes