accessibility menu, dialog, popup

UserName

H3COHCH3

CAS RN 589-82-2

IUPAC Name: heptan-3-ol
Synonyms: 3-heptanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: RZKSECIXORKHQS-UHFFFAOYNA-N
SMILES: CCCCC(O)CC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)