missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanol 98.0+%, TCI America™

Catalog No. H0035500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0035500ML 500 mL
H003525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0035500ML Supplier TCI America Supplier No. H0035500ML

Chemical Identifiers

CAS 589-82-2
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00004586
InChI Key RZKSECIXORKHQS-UHFFFAOYSA-N
Synonym 3-heptanol, 3-hydroxyheptane, ethyl butyl carbinol, butyl ethyl carbinol, butylethylcarbinol, xi-3-heptanol, fema no. 3547, ethylbutylcarbinol, heptanol-3, 3-heptyl alcohol
PubChem CID 11520
IUPAC Name heptan-3-ol
SMILES CCCCC(CC)O

Specifications

Color Colorless
Boiling Point 156°C
Quantity 500 mL
UN Number 1987
Formula Weight 116.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Heptanol
EINECSNumber (2)-0217
RTECSNumber MJ3150000
TSCA Yes