missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Glycidyloxypropyltrimethoxysilane 97.0+%, TCI America™
Supplier: TCI America G021025G
Specifications
| 3-Glycidyloxypropyltrimethoxysilane | |
| Colorless | |
| MFCD00005144 | |
| 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane | |
| CO[Si](CCCOCC1CO1)(OC)OC | |
| 236.339 | |
| 236.34 | |
| Liquid |
| 2530-83-8 | |
| C9H20O5Si | |
| 25 g | |
| BPSIOYPQMFLKFR-UHFFFAOYSA-N | |
| trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane | |
| 17317 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 2530-83-8 | |
| 236.339 | |
| BPSIOYPQMFLKFR-UHFFFAOYSA-N | |
| 17317 | |
| CO[Si](CCCOCC1CO1)(OC)OC |
| C9H20O5Si | |
| MFCD00005144 | |
| 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane | |
| trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane |
Safety and Handling
EINECSNumber : (2)-2071
RTECSNumber : VV4025000
TSCA : Yes