missing translation for 'onlineSavingsMsg'
Learn More

3-Glycidyloxypropyltrimethoxysilane 97.0+%, TCI America™

Catalog No. p-7130191
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G021025G 25 g
G0210100G 100 g
G0210500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G021025G Supplier TCI America Supplier No. G021025G
Only null left

Chemical Identifiers

CAS 2530-83-8
Molecular Formula C9H20O5Si
Molecular Weight (g/mol) 236.339
MDL Number MFCD00005144
InChI Key BPSIOYPQMFLKFR-UHFFFAOYSA-N
Synonym 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
PubChem CID 17317
IUPAC Name trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane
SMILES CO[Si](CCCOCC1CO1)(OC)OC

Specifications

Color Colorless
Quantity 25 g
Formula Weight 236.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Glycidyloxypropyltrimethoxysilane
EINECSNumber (2)-2071
RTECSNumber VV4025000
TSCA Yes