missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Furancarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America F030425G
Specifications
| 3-Furancarboxylic Acid | |
| 122°C | |
| MFCD00005350 | |
| 3-furoic acid, 3-furancarboxylic acid, 3-carboxyfuran, 3-furanoic acid, unii-88a3s7rg98, 3-furoate, 3-furans, 3-furanoate, pubchem6945, acmc-209kee | |
| OC(=O)C1=COC=C1 | |
| 112.08 | |
| CHEBI:30846 | |
| ≥98.0% (T) |
| 488-93-7 | |
| C5H4O3 | |
| 25 g | |
| IHCCAYCGZOLTEU-UHFFFAOYSA-N | |
| furan-3-carboxylic acid | |
| 10268 | |
| 112.08 | |
| Crystalline Powder |
Chemical Identifiers
| 488-93-7 | |
| 112.08 | |
| IHCCAYCGZOLTEU-UHFFFAOYSA-N | |
| 10268 | |
| furan-3-carboxylic acid |
| C5H4O3 | |
| MFCD00005350 | |
| 3-furoic acid, 3-furancarboxylic acid, 3-carboxyfuran, 3-furanoic acid, unii-88a3s7rg98, 3-furoate, 3-furans, 3-furanoate, pubchem6945, acmc-209kee | |
| CHEBI:30846 | |
| OC(=O)C1=COC=C1 |
Safety and Handling
TSCA : No