missing translation for 'onlineSavingsMsg'
Learn More

3-Furancarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129602
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03045G 5 g
F030425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03045G Supplier TCI America Supplier No. F03045G
Only null left

Chemical Identifiers

CAS 488-93-7
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005350
InChI Key IHCCAYCGZOLTEU-UHFFFAOYSA-N
Synonym 3-furoic acid, 3-furancarboxylic acid, 3-carboxyfuran, 3-furanoic acid, unii-88a3s7rg98, 3-furoate, 3-furans, 3-furanoate, pubchem6945, acmc-209kee
PubChem CID 10268
ChEBI CHEBI:30846
IUPAC Name furan-3-carboxylic acid
SMILES OC(=O)C1=COC=C1

Specifications

Melting Point 122°C
Quantity 5 g
Formula Weight 112.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Furancarboxylic Acid