missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Furaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
$115.10 - $115.10
Chemical Identifiers
| CAS | 216144-29-5 |
|---|---|
| Molecular Formula | C9H14O3 |
| Molecular Weight (g/mol) | 170.21 |
| MDL Number | MFCD01318151 |
| InChI Key | MTZMXGCDOYBCPN-UHFFFAOYSA-N |
| Synonym | 3-diethoxymethyl furan, 3-furaldehyde diethyl acetal, furan,3-diethoxymethyl, diethoxy-3-furylmethane, acmc-1ckie |
| PubChem CID | 7006657 |
| IUPAC Name | 3-(diethoxymethyl)furan |
| SMILES | CCOC(OCC)C1=COC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1551906
|
Thermo Scientific Chemicals
L1551906 |
5 g |
Each for $115.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 216144-29-5 | |
| 170.21 | |
| MTZMXGCDOYBCPN-UHFFFAOYSA-N | |
| 7006657 | |
| CCOC(OCC)C1=COC=C1 |
| C9H14O3 | |
| MFCD01318151 | |
| 3-diethoxymethyl furan, 3-furaldehyde diethyl acetal, furan,3-diethoxymethyl, diethoxy-3-furylmethane, acmc-1ckie | |
| 3-(diethoxymethyl)furan |
Specifications
| 216144-29-5 | |
| 1.444 | |
| 5 g | |
| MTZMXGCDOYBCPN-UHFFFAOYSA-N | |
| 3-(diethoxymethyl)furan | |
| 7006657 | |
| 98% |
| C9H14O3 | |
| MFCD01318151 | |
| 3-diethoxymethyl furan, 3-furaldehyde diethyl acetal, furan,3-diethoxymethyl, diethoxy-3-furylmethane, acmc-1ckie | |
| CCOC(OCC)C1=COC=C1 | |
| 170.21 | |
| 170.21 | |
| 3-Furaldehyde diethyl acetal |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only