missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Furaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1551906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Furaldehyde diethyl acetal | |
| C9H14O3 | |
| MFCD01318151 | |
| 3-diethoxymethyl furan, 3-furaldehyde diethyl acetal, furan,3-diethoxymethyl, diethoxy-3-furylmethane, acmc-1ckie | |
| CCOC(OCC)C1=COC=C1 | |
| 170.21 | |
| 170.21 |
| 216144-29-5 | |
| 1.444 | |
| 5 g | |
| MTZMXGCDOYBCPN-UHFFFAOYSA-N | |
| 3-(diethoxymethyl)furan | |
| 7006657 | |
| 98% |
Chemical Identifiers
| 216144-29-5 | |
| 170.21 | |
| MTZMXGCDOYBCPN-UHFFFAOYSA-N | |
| 7006657 | |
| CCOC(OCC)C1=COC=C1 |
| C9H14O3 | |
| MFCD01318151 | |
| 3-diethoxymethyl furan, 3-furaldehyde diethyl acetal, furan,3-diethoxymethyl, diethoxy-3-furylmethane, acmc-1ckie | |
| 3-(diethoxymethyl)furan |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only