missing translation for 'onlineSavingsMsg'
Learn More

3-Formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F04451G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04451G Supplier TCI America Supplier No. F04451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87199-16-4
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00161356
InChI Key HJBGZJMKTOMQRR-UHFFFAOYSA-N
Synonym 3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
PubChem CID 2734356
IUPAC Name (3-formylphenyl)boronic acid
SMILES OB(O)C1=CC=CC(C=O)=C1

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 1 g
Formula Weight 149.94
Physical Form Crystalline Powder
Chemical Name or Material 3-Formylphenylboronic Acid (contains varying amounts of Anhydride)