missing translation for 'onlineSavingsMsg'
Learn More

3-Formylphenylboronic acid, 97%

CAS: 87199-16-4 | C7H7BO3 | 149.94 g/mol

$178.75 - $178.75

Chemical Identifiers

CAS 87199-16-4
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00161356
InChI Key HJBGZJMKTOMQRR-UHFFFAOYSA-N
Synonym 3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
PubChem CID 2734356
IUPAC Name (3-formylphenyl)boronic acid
SMILES OB(O)C1=CC=CC(C=O)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359370050
SDS View Documents
Thermo Scientific Chemicals
359370050
5 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

87199-16-4
149.94
HJBGZJMKTOMQRR-UHFFFAOYSA-N
2734356
OB(O)C1=CC=CC(C=O)=C1
C7H7BO3
MFCD00161356
3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
(3-formylphenyl)boronic acid

Specifications

87199-16-4
100.0
Beige to White or Orange
96% min. (HPLC)
C7H7BO3
5 g
HJBGZJMKTOMQRR-UHFFFAOYSA-N
(3-formylphenyl)boronic acid
2734356
97%
3-Formylphenylboronic acid
96.0
100°C
Authentic
Glass bottle
MFCD00161356
3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
OB(O)C1=CC=CC(C=O)=C1
149.94
149.94
Powder, Crystals or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only