missing translation for 'onlineSavingsMsg'
Learn More

3-Formylbenzeneboronic acid

CAS: 87199-16-4 | C7H7BO3 | 149.94 g/mol

$59.85 - $658.47

Chemical Identifiers

CAS 87199-16-4
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00161356
InChI Key HJBGZJMKTOMQRR-UHFFFAOYSA-N
Synonym 3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
PubChem CID 2734356
IUPAC Name (3-formylphenyl)boronic acid
SMILES OB(O)C1=CC=CC(C=O)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2543703
View Documents
Thermo Scientific Chemicals
B2543703
1 g
Each for $59.85
Only null left
 
AAB2543706
View Documents
Thermo Scientific Chemicals
B2543706
5 g
Each for $180.05
Only null left
 
AAB2543714
View Documents
Thermo Scientific Chemicals
B2543714
25 g
Each for $658.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

87199-16-4
149.94
HJBGZJMKTOMQRR-UHFFFAOYSA-N
2734356
OB(O)C1=CC=CC(C=O)=C1
C7H7BO3
MFCD00161356
3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
(3-formylphenyl)boronic acid

Specifications

87199-16-4
C7H7BO3
1 g
Air Sensitive
HJBGZJMKTOMQRR-UHFFFAOYSA-N
(3-formylphenyl)boronic acid
2734356
3-Formylbenzeneboronic acid
109°C to 113°C
MFCD00161356
3030769
3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
OB(O)C1=CC=CC(C=O)=C1
149.94
149.94
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only