missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorotoluene 99.0+%, TCI America™

Catalog No. F004025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F004025ML 25 mL
F0040500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F004025ML Supplier TCI America Supplier No. F004025ML

Chemical Identifiers

CAS 352-70-5
Molecular Formula C7H7F
Molecular Weight (g/mol) 110.13
MDL Number MFCD00000339
InChI Key BTQZKHUEUDPRST-UHFFFAOYSA-N
Synonym 3-fluorotoluene, m-fluorotoluene, benzene, 1-fluoro-3-methyl, 1-fluoro-3-methyl-benzene, 1-methyl-3-fluorobenzene, toluene, m-fluoro, unii-f72w445dq0, 3-fluorobenzyl radical, meta-fluorotoluene, acmc-209ifn
PubChem CID 9606
IUPAC Name 1-fluoro-3-methylbenzene
SMILES CC1=CC=CC(F)=C1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 mL
UN Number 2388
Formula Weight 110.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorotoluene
EINECSNumber (3)-0083
TSCA Yes