missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorotoluene, 98%

Catalog No. p-7023054
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1610714 25 g
AAA1610722 100 g
AAA1610730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1610714 Supplier Thermo Scientific Chemicals Supplier No. A1610714
Only null left

CAS: 352-70-5 | C7H7F | 110.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 352-70-5
Molecular Formula C7H7F
Molecular Weight (g/mol) 110.13
MDL Number MFCD00000339
InChI Key BTQZKHUEUDPRST-UHFFFAOYSA-N
Synonym 3-fluorotoluene, m-fluorotoluene, benzene, 1-fluoro-3-methyl, 1-fluoro-3-methyl-benzene, 1-methyl-3-fluorobenzene, toluene, m-fluoro, unii-f72w445dq0, 3-fluorobenzyl radical, meta-fluorotoluene, acmc-209ifn
PubChem CID 9606
IUPAC Name 1-fluoro-3-methylbenzene
SMILES CC1=CC=CC(F)=C1

Specifications

Melting Point -87°C
Density 0.993
Boiling Point 114°C to 116°C
Flash Point 9°C (48°F)
Odor Aromatic
Refractive Index 1.469
Quantity 25 g
UN Number UN2388
Beilstein 1903631
Merck Index 14,4180
Formula Weight 110.13
Percent Purity 98%
Chemical Name or Material 3-Fluorotoluene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H335
Highly flammable liquid and vapour.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES
EINECSNumber 206-524-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only