missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorothiophene 98.0+%, TCI America™
Supplier: TCI America F05121G
Specifications
| 3-Fluorothiophene | |
| Yellow | |
| C4H3FS | |
| 1 g | |
| WPAQIMRFMFRJTP-UHFFFAOYSA-N | |
| 3-fluorothiophene | |
| 10975421 | |
| ≥98.0% (GC) |
| 3093-88-7 | |
| 85°C | |
| MFCD08275189 | |
| 1993 | |
| C1=CSC=C1F | |
| 102.126 | |
| 102.13 | |
| Liquid |
Chemical Identifiers
| 3093-88-7 | |
| 102.126 | |
| WPAQIMRFMFRJTP-UHFFFAOYSA-N | |
| 3-fluorothiophene |
| C4H3FS | |
| MFCD08275189 | |
| 10975421 | |
| C1=CSC=C1F |
Safety and Handling
TSCA : No
Recommended Storage : Freezer