missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorothiophene 98.0+%, TCI America™

Catalog No. p-7129721
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0512200MG 200 mg
F05121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0512200MG Supplier TCI America Supplier No. F0512200MG

Chemical Identifiers

CAS 3093-88-7
Molecular Formula C4H3FS
Molecular Weight (g/mol) 102.126
MDL Number MFCD08275189
InChI Key WPAQIMRFMFRJTP-UHFFFAOYSA-N
PubChem CID 10975421
IUPAC Name 3-fluorothiophene
SMILES C1=CSC=C1F

Specifications

Color Yellow
Boiling Point 85°C
Quantity 200 mg
UN Number 1993
Formula Weight 102.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorothiophene
TSCA No
Recommended Storage Freezer