missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine 98.0+%, TCI America™

Supplier:  TCI America F020725G

 View more versions of this product

Catalog No. F020725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Fluoropyridine
Yellow
C5H4FN
25 g
pyridine, 3-fluoro, 3-fluorpyridin, ccris 1717, 3-fluropyridine, 3-fluoro-pyridine, 3-fluoro-pyridinium, pubchem1095, acmc-1coty, 3-fluoropyridine, ksc173o1l
C1=CC(=CN=C1)F
97.092
97.09
Liquid
372-47-4
106°C
MFCD00006374
1993
CELKOWQJPVJKIL-UHFFFAOYSA-N
3-fluoropyridine
67794
≥98.0% (GC)

Chemical Identifiers

372-47-4
97.092
CELKOWQJPVJKIL-UHFFFAOYSA-N
67794
C1=CC(=CN=C1)F
C5H4FN
MFCD00006374
pyridine, 3-fluoro, 3-fluorpyridin, ccris 1717, 3-fluropyridine, 3-fluoro-pyridine, 3-fluoro-pyridinium, pubchem1095, acmc-1coty, 3-fluoropyridine, ksc173o1l
3-fluoropyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions