missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine 98.0+%, TCI America™

Catalog No. p-7129539
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02075G 5 g
F020725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02075G Supplier TCI America Supplier No. F02075G
Only null left

Chemical Identifiers

CAS 372-47-4
Molecular Formula C5H4FN
Molecular Weight (g/mol) 97.092
MDL Number MFCD00006374
InChI Key CELKOWQJPVJKIL-UHFFFAOYSA-N
Synonym pyridine, 3-fluoro, 3-fluorpyridin, ccris 1717, 3-fluropyridine, 3-fluoro-pyridine, 3-fluoro-pyridinium, pubchem1095, acmc-1coty, 3-fluoropyridine, ksc173o1l
PubChem CID 67794
IUPAC Name 3-fluoropyridine
SMILES C1=CC(=CN=C1)F

Specifications

Color Yellow
Boiling Point 106°C
Quantity 5 g
UN Number 1993
Formula Weight 97.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluoropyridine