missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine-4-boronic acid, 98%

Catalog No. p-7035392
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53109MD 250 mg
AAH5310903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53109MD Supplier Thermo Scientific Chemicals Supplier No. H53109MD
Only null left

CAS: 458532-97-3 | C5H5BFNO2 | 140.908 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 458532-97-3
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.908
MDL Number MFCD03788558
InChI Key QUHSESYLMZVXCN-UHFFFAOYSA-N
Synonym 3-fluoropyridine-4-boronic acid, 3-fluoropyridin-4-yl boronic acid, 3-fluoropyridin-4-yl-4-boronic acid, 3-fluoro-4-pyridyl boronic acid, boronic acid, 3-fluoro-4-pyridinyl, 3-fluoro-4-pyridylboronic acid, 3-fluoropyridin-4-yl boranediol, 3-fluoro-4-pyridineboronic acid, 3-fluoropyridine-4-boronicacid
PubChem CID 2779353
IUPAC Name (3-fluoropyridin-4-yl)boronic acid
SMILES B(C1=C(C=NC=C1)F)(O)O

Specifications

Melting Point 237°C to 239°C
Quantity 250 mg
Formula Weight 140.91
Percent Purity 98%
Chemical Name or Material 3-Fluoropyridine-4-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only