accessibility menu, dialog, popup

UserName

HOBOHNF

CAS RN 458532-97-3

IUPAC Name: (3-fluoropyridin-4-yl)boronic acid
Synonyms: 3-fluoropyridin-4-ylboronic acid
Molecular Weight (g/mol): 140.91
Molecular Formula: C5H5BFNO2
InChi Key: QUHSESYLMZVXCN-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=NC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results