missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophthalic Acid 98.0+%, TCI America™

Catalog No. F03535G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03535G Supplier TCI America Supplier No. F03535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1583-67-1
Molecular Formula C8H5FO4
Molecular Weight (g/mol) 184.122
MDL Number MFCD00039524
InChI Key BBCQSMSCEJBIRD-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1,2-dicarboxylic acid, 3-fluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylic acid, 3-fluoro, 1,2-benzenedicarboxylic acid, fluoro, 3-fluorophthalic, 3-fluorophtalic acid, 3-fluorophthatic acid, pubchem1405, 3-fluorophthalicacid
PubChem CID 345381
IUPAC Name 3-fluorophthalic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)C(=O)O

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
Formula Weight 184.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorophthalic Acid