missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylmagnesium bromide, 1M solution in THF, AcroSeal™

Catalog No. AC427821000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC427821000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427821000 Supplier Thermo Scientific Chemicals Supplier No. 427821000
Only null left

CAS: 17318-03-5 | C6H4BrFMg | 199.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17318-03-5 , 109-99-9
Molecular Formula C6H4BrFMg
Molecular Weight (g/mol) 199.31
MDL Number MFCD00672007
InChI Key VFPDAAQAKDGSHQ-UHFFFAOYSA-M
Synonym 3-fluorophenylmagnesium bromide, bromo 3-fluorophenyl magnesium, 3-flurophenylmagnesiumbromide, 3-fluorophenylmagnesiumbromide, 3-flurophenyl magnesium bromide, 3-fluorophenyl magnesium bromide, 3-fluorophenyl-magnesium bromide, vfpdaaqakdgshq-uhfffaoysa-m, 3-fluorophenylmagnesium bromide, 1m in methf, grignard reagent
PubChem CID 2778196
IUPAC Name bromo(3-fluorophenyl)magnesium
SMILES FC1=CC([Mg]Br)=CC=C1

Specifications

Color Yellow to brown solution
Boiling Point 65.0°C
CAS Min % 19.46, 78.59
CAS Max % 21.41, 80.54
Concentration 1M (Complexometry), exact strength on the certificate of analysis
Density 1.0240g/mL
Formula Weight 199.3
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.024
Quantity 100 mL
Flash Point −21°C
Chemical Name or Material 3-Fluorophenylmagnesium bromide, 1M solution in THF
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Suspected of causing cancer.
In contact with water releases flammable gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only