missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorophenylacetic Acid 98.0+%, TCI America™
Supplier: TCI America F021625G
Specifications
| 3-Fluorophenylacetic Acid | |
| 47°C | |
| 125°C | |
| MFCD00004331 | |
| 3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid | |
| OC(=O)CC1=CC=CC(F)=C1 | |
| 154.14 | |
| 154.14 | |
| Crystalline Powder |
| 331-25-9 | |
| White | |
| C8H7FO2 | |
| 25 g | |
| YEAUYVGUXSZCFI-UHFFFAOYSA-N | |
| 2-(3-fluorophenyl)acetic acid | |
| 67617 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 331-25-9 | |
| 154.14 | |
| YEAUYVGUXSZCFI-UHFFFAOYSA-N | |
| 67617 | |
| OC(=O)CC1=CC=CC(F)=C1 |
| C8H7FO2 | |
| MFCD00004331 | |
| 3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid | |
| 2-(3-fluorophenyl)acetic acid |
Safety and Handling
TSCA : Yes